Skip to Content
Merck
All Photos(1)

Documents

428450

Sigma-Aldrich

2,4,6-Trimethylbenzyl mercaptan

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2CH2SH
CAS Number:
Molecular Weight:
166.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

120 °C/5 mmHg (lit.)

mp

46-48 °C (lit.)

SMILES string

Cc1cc(C)c(CS)c(C)c1

InChI

1S/C10H14S/c1-7-4-8(2)10(6-11)9(3)5-7/h4-5,11H,6H2,1-3H3

InChI key

LJZYEPXPMRRXTA-UHFFFAOYSA-N

General description

2,4,6-Trimethylbenzyl mercaptan is an aromatic mercaptan derivative. Its physical properties like vapor pressure, density, freezing point, boiling point, refractive index and enthalpy of vaporization at boiling point has been determined.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carl L. Yaws
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 292-292 (2015)
Carl L. Yaws
The Yaws Handbook of Vapor Pressure: Antoine coefficients , 97-97 (2015)
Carl L. Yaws
Thermophysical Properties of Chemicals and Hydrocarbons, 582-582 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service