Skip to Content
Merck
All Photos(1)

Documents

425591

Sigma-Aldrich

4,6-Dichloro-2,5-diphenylpyrimidine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10Cl2N2
CAS Number:
Molecular Weight:
301.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

130-133 °C (lit.)

SMILES string

Clc1nc(nc(Cl)c1-c2ccccc2)-c3ccccc3

InChI

1S/C16H10Cl2N2/c17-14-13(11-7-3-1-4-8-11)15(18)20-16(19-14)12-9-5-2-6-10-12/h1-10H

InChI key

SYZDNTMHIDTVBR-UHFFFAOYSA-N

General description

4,6-Dichloro-2,5-diphenylpyrimidine is a nucleobase derivative.

Application

4,6-Dichloro-2,5-diphenylpyrimidine has been used as a test compound in the systematic quantitative structure-retention relationship (QSRR) studies between experimental chromatographic retention data and molecular descriptors. It may be used as an electrophile in the synthesis of H-chelate containing 2-quinazolylacetonitrile derivatives by reacting with 2-cyanomethylquinazoline.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of new 2-quinazolylacetonitrile derivatives.
Kim JH and Min KS.
J. Korean Chem. Soc., 45(6), 532-537 (2001)
Multivariate Chemometric Study on the Interfacial Properties of Nucleic-Acid Bases.
Luo HB, et al.
Journal of Data Science, 1(4), 447-460 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service