Skip to Content
Merck
All Photos(1)

Key Documents

421448

Sigma-Aldrich

Zonyl® FSO fluorosurfactant

Synonym(s):

nonionic ethoxylated fluorosurfactant

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)x(CF2)yC2H5FO
CAS Number:
UNSPSC Code:
12162002

refractive index

n20/D 1.3910

bp

100 °C

density

1.3 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Zonyl is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service