Skip to Content
Merck
All Photos(1)

Documents

408697

Sigma-Aldrich

(R)-(−)-2-Heptanol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH3
CAS Number:
Molecular Weight:
116.20
Beilstein:
1719090
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

95%

optical activity

[α]20/D −9.5°, neat

refractive index

n20/D 1.419 (lit.)

bp

74-75 °C/23 mmHg (lit.)

density

0.818 g/mL at 25 °C (lit.)

SMILES string

CCCCC[C@@H](C)O

InChI

1S/C7H16O/c1-3-4-5-6-7(2)8/h7-8H,3-6H2,1-2H3/t7-/m1/s1

InChI key

CETWDUZRCINIHU-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service