Skip to Content
Merck
All Photos(1)

Documents

40653

Sigma-Aldrich

(R)-7,7′-Bis[di(3,5-xylyl)phosphino]-3,3′,4,4′-tetrahydro-4,4′-dimethyl-8,8′-bi(2H-1,4-benzoxazine)

≥97%

Synonym(s):

(R)-7,7′-Bis[bis(3,5-dimethylphenyl)phosphino]-3,3′,4,4′-tetrahydro-4,4′-dimethyl-8,8′-bi(2H-1,4-benzoxazine), Xylyl-Solphos SL-A002-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H54N2O2P2
CAS Number:
Molecular Weight:
776.92
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥97%

optical purity

ee: ≥99%

SMILES string

CN1CCOc2c1ccc(P(c3ccccc3)c4ccccc4)c2-c5c6OCCN(C)c6ccc5P(c7ccccc7)c8ccccc8

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service