Skip to Content
Merck
All Photos(1)

Key Documents

406252

Sigma-Aldrich

Boc-β-iodo-Ala-OBzl

97%

Synonym(s):

N-(tert-Butoxycarbonyl)-3-iodo-L-alanine benzyl ester, Boc-3-iodo-L-alanine benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2CH[NHCO2C(CH3)3]CO2CH2C6H5
CAS Number:
Molecular Weight:
405.23
Beilstein:
4506658
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D −18.0°, c = 1 in ethanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

78-80 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](CI)C(=O)OCc1ccccc1

InChI

1S/C15H20INO4/c1-15(2,3)21-14(19)17-12(9-16)13(18)20-10-11-7-5-4-6-8-11/h4-8,12H,9-10H2,1-3H3,(H,17,19)/t12-/m0/s1

InChI key

DDXFSYLOWHQCEK-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service