Skip to Content
Merck
All Photos(1)

Documents

403695

Sigma-Aldrich

Neodymium(III) titanate

−325 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2Nd2O3 · 5TiO2
Molecular Weight:
1072.29
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

reaction suitability

reagent type: catalyst
core: neodymium

particle size

−325 mesh

SMILES string

O=[Ti]=O.O=[Ti]=O.O=[Ti]=O.O=[Ti]=O.O=[Nd]O[Nd]=O.O=[Nd]O[Nd]=O

InChI

1S/4Nd.14O.4Ti

InChI key

XXBVDTWSFRCPKR-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service