Skip to Content
Merck
All Photos(1)

Key Documents

394971

Supelco

N,N-Dimethylformamide diethyl acetal

for GC derivatization

Synonym(s):

1,1-Diethoxy-N,N-dimethylmethylamine, 1,1-Diethoxytrimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH(OC2H5)2
CAS Number:
Molecular Weight:
147.22
Beilstein:
741889
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

for GC derivatization

reaction suitability

reagent type: derivatization reagent
reaction type: Acetylations

refractive index

n20/D 1.400 (lit.)

bp

130-133 °C (lit.)

density

0.859 g/mL at 25 °C (lit.)

SMILES string

CCOC(OCC)N(C)C

InChI

1S/C7H17NO2/c1-5-9-7(8(3)4)10-6-2/h7H,5-6H2,1-4H3

InChI key

BWKAYBPLDRWMCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for the derivatization of primary sulfonamides.

Other Notes

Reagent for n-dimethylaminomethylene.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.R. Knapp
Handbook of Analytical Derivatization Reactions, 398-398 (1979)
W.J.A. VandenHeuval and V.F. Gruber
Journal of Chromatography A, 112, 513-513 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service