Skip to Content
Merck
All Photos(1)

Documents

393819

Sigma-Aldrich

4-Acetylphenoxyacetic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COC6H4OCH2CO2H
CAS Number:
Molecular Weight:
194.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

175-177 °C (dec.) (lit.)

solubility

methanol: soluble 25 mg/mL, clear, colorless to faintly yellow

SMILES string

CC(=O)c1ccc(OCC(O)=O)cc1

InChI

1S/C10H10O4/c1-7(11)8-2-4-9(5-3-8)14-6-10(12)13/h2-5H,6H2,1H3,(H,12,13)

InChI key

KMXZEXUYXUMHEQ-UHFFFAOYSA-N

General description

4-Acetylphenoxyacetic acid is a phenoxyacetic acid derivative. It has been synthesized by employing para-acetyl phenol as starting reagent.

Application

4-Acetylphenoxyacetic acid is suitable as a test compound in the study of phenylacetic acid transport system (PATS) of Pseudomonas putida U by the aerobic catabolism of phenylacetic acid derivative used as the sole carbon source.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Schleissner et al.
Journal of bacteriology, 176(24), 7667-7676 (1994-12-01)
The phenylacetic acid transport system (PATS) of Pseudomonas putida U was studied after this bacterium was cultured in a chemically defined medium containing phenylacetic acid (PA) as the sole carbon source. Kinetic measurement was carried out, in vivo, at 30
Photocyclization reactions. Part 1. Synthesis of dihydrobenzofuranols using photocyclization of 2-alkoxybenzaldehydes, 2'-alkoxyacetophenones, 2-formylphenoxyacetic acids and 2-acetylphenoxyacetic acids
Horaguchi T, et al.
Journal of Heterocyclic Chemistry, 28(5), 1261-1272 (1991)
Design of new potent hypolipidemic agents with the synergistic structural properties of α-asarone and fibrates.
Zuniga C, et al.
Drug Development Research, 64(1), 28-40 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service