Skip to Content
Merck
All Photos(1)

Key Documents

385581

Sigma-Aldrich

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline

98%

Synonym(s):

2,3,5,6,α,α,α-Heptafluoro-p-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6F4NH2
CAS Number:
Molecular Weight:
233.09
Beilstein:
2657893
MDL number:
UNSPSC Code:
12352100

Assay

98%

refractive index

n20/D 1.431 (lit.)

bp

186 °C (lit.)

density

1.687 g/mL at 25 °C (lit.)

SMILES string

Nc1c(F)c(F)c(c(F)c1F)C(F)(F)F

InChI

1S/C7H2F7N/c8-2-1(7(12,13)14)3(9)5(11)6(15)4(2)10/h15H2

InChI key

FJOACTZFMHZHSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service