Skip to Content
Merck
All Photos(1)

Key Documents

369578

Sigma-Aldrich

4-Methylphenyl benzoate

99%

Synonym(s):

p-Tolyl benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2C6H4CH3
CAS Number:
Molecular Weight:
212.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

solid

mp

70-72 °C (lit.)

functional group

ester
phenyl

SMILES string

Cc1ccc(OC(=O)c2ccccc2)cc1

InChI

1S/C14H12O2/c1-11-7-9-13(10-8-11)16-14(15)12-5-3-2-4-6-12/h2-10H,1H3

InChI key

LLRZUDIHEZXFGV-UHFFFAOYSA-N

General description

4-Methylphenyl benzoate is an ester. Structure of 4-methylphenyl benzoate resembles phenyl benzoate and 4-methoxy­phenyl benzoate, in terms of similar geometric parameters. The dihedral angle between the phenyl and benzene rings in 4-methylphenyl benzoate is 60.17 (7)°.[1] Fries rearrangement of 4-methoxy­phenyl benzoate catalyzed by sulfated zirconia is reported.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genuinely catalytic Fries rearrangement using sulfated zirconia.
Clark JH, et al.
Green Chemistry, 4(4), 366-368 (2002)
4-Methylphenyl benzoate.
Gowda BT, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(8), o3563-o3563 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service