Skip to Content
Merck
All Photos(1)

Key Documents

36612

Supelco

4-Nitrophenol sodium salt dihydrate

PESTANAL®, analytical standard

Synonym(s):

Sodium 4-nitrophenoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4ONa · 2H2O
CAS Number:
Molecular Weight:
197.12
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

mp

>300 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

O.O.[Na+].[O-]c1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H5NO3.Na.2H2O/c8-6-3-1-5(2-4-6)7(9)10;;;/h1-4,8H;;2*1H2/q;+1;;/p-1

InChI key

ZJYUDTPVWWBKPV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service