Skip to Content
Merck
All Photos(1)

Documents

365769

Sigma-Aldrich

2-Bromo-5-nitrobenzotrifluoride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(NO2)CF3
CAS Number:
Molecular Weight:
270.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

87-88 °C/3 mmHg (lit.)

mp

41-44 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(Br)c(c1)C(F)(F)F

InChI

1S/C7H3BrF3NO2/c8-6-2-1-4(12(13)14)3-5(6)7(9,10)11/h1-3H

InChI key

SXEQQBBOAMHOID-UHFFFAOYSA-N

Application

2-Bromo-5-nitrobenzotrifluoride may be used in the preparation of:
  • new unsymmetrical diamine monomer containing both the benzimidazole ring and trifluoromethyl group, 6,4′-diamino-2′-trifluoromethyl-2-phenylbenzimidazole
  • 2,5-bis[(4-nitro-2-trifluoromethylphenoxy)-phenyl]diphenylphosphine oxide (BNTFDPO)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organo-soluble and transparent polyimides containing phenylphosphine oxide and trifluoromethyl moiety: Synthesis and characterization.
Li Z, et al.
Eur. Polymer J., 45(4), 1139-1148 (2009)
Soluble polyimides from unsymmetrical diamine containing benzimidazole ring and trifluoromethyl pendent group.
Choi H, et al.
Polymer, 49(11), 2644-2649 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service