Skip to Content
Merck
All Photos(1)

Key Documents

365602

Sigma-Aldrich

Sinomenine

0.3 mol chloroform of crystallization

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO4 · 0.3CHCl3
CAS Number:
Molecular Weight:
365.20
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

impurities

0.3 mol chloroform of crystallization

mp

180 °C (dec.) (lit.)

functional group

ether
ketone

SMILES string

COC1=CC2[C@@H]3Cc4ccc(OC)c(O)c4[C@]2(CCN3C)CC1=O

InChI

1S/C19H23NO4/c1-20-7-6-19-10-14(21)16(24-3)9-12(19)13(20)8-11-4-5-15(23-2)18(22)17(11)19/h4-5,9,12-13,22H,6-8,10H2,1-3H3/t12-,13+,19-/m1/s1

InChI key

INYYVPJSBIVGPH-QHRIQVFBSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng-Xiang Su et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 889-890, 39-43 (2012-02-22)
A sensitive and selective liquid chromatographic tandem mass spectrometric method was developed and validated for the determination of sinomenine in human plasma. Plasma samples were precipitated using methanol with metronidazole as internal standard. Separation was carried out on an Inertsil
Mei-Yu Zhang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 38(4), 597-604 (2013-05-30)
To observe the analgesic effect of sinomenine on the neuropathic pain rat model induced by SSNI, and discuss its impact on monoamine neurotransmitters in striatal extracellular fluid. Male SD rats were randomly divided into the sham operation group, the SSNI
Peng Teng et al.
European journal of medicinal chemistry, 50, 63-74 (2012-02-14)
Sinomenine (1) is clinically available for the treatment of rheumatoid arthritis (RA), however, its efficacy is quite weak. In the present study, a library of novel sinomenine-based homodimers and monomers through variable-length linkers were designed and synthesized, and their bioactivities
Yi Hong et al.
Cancer immunology, immunotherapy : CII, 62(3), 447-454 (2012-09-04)
Transferrin receptor (TfR) has been used as a target for the antibody-based therapy of cancer due to its higher expression in tumors relative to normal tissues. Great potential has been shown by anti-TfR antibodies combined with chemotherapeutic drugs as a
Y C Oh et al.
European review for medical and pharmacological sciences, 16(9), 1184-1191 (2012-10-11)
Sinomenine is an alkaloid compound and a prominent anti-inflammatory agent found in the root of the climbing plant Sinomenium acutum. However, its effects on the mechanism of human mast cell line (HMC)-1-mediated inflammation remained unknown. To provide insight into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service