Skip to Content
Merck
All Photos(2)

Key Documents

363308

Sigma-Aldrich

Carbostyril 124

99%

Synonym(s):

7-Amino-4-methyl-2(1H)-quinolinone, 7-Amino-4-methyl-2-hydroxyquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2O
CAS Number:
Molecular Weight:
174.20
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99%

form

solid

mp

278-280 °C (lit.)

λmax

350 nm

SMILES string

CC1=CC(=O)Nc2cc(N)ccc12

InChI

1S/C10H10N2O/c1-6-4-10(13)12-9-5-7(11)2-3-8(6)9/h2-5H,11H2,1H3,(H,12,13)

InChI key

MJXYFLJHTUSJGU-UHFFFAOYSA-N

General description

Carbostyril 124 (CS124) is a chromophore that can be used as a laser dye for sensitization and optical detection.[1][2] It acts as an antenna molecule that facilitates the light absorption and enhances fluorescence efficiency.[1][2][3]

Application

CS124 can be used in the synthesis of luminescent terbium chelates (LTCs) to develop fluoroimmunoassay for the detection of carcinoembryonic antigen.[4] It can also be used in the development of borane lasers.[5]
Suitable as a laser dye

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A borane laser
Cerdan L, et al.
Nature Communications, 6(4), 5958-5958 (2015)
A novel homogeneous time-resolved fluoroimmunoassay for carcinoembryonic antigen based on water-soluble quantum dots
Chen Z, et al.
Journal of Fluorescence, 23(4), 649-657 (2013)
Subnanosecond light pulses from GaAs injection lasers.
Lee T and Roldan R
IEEE Journal of Quantum Electronics, 5(11), 551-552 (1969)
G Kokotos et al.
Biological chemistry Hoppe-Seyler, 371(9), 835-840 (1990-09-01)
Sensitive and convenient fluorometric assays for the determination of chymotrypsin have been developed by using the substrates Glt-Leu-Phe-NH-Meq, Glt-Phe-NH-FMeq and Glt-Leu-Phe-NH-FMeq, which have been synthesized utilizing the mixed anhydride method. The amidolytic activity of chymotrypsin was measured by the release
Modest effectiveness of carbostyril 124 as a sensitising chromophore in europium and terbium amide complexes based on 1, 4, 7, 10-tetraazacyclododecane.
Parker D and Williams JAG
J. Chem. Soc. Perkin Trans. II, 1581-1586 (1996)

Articles

Developed in the last several years, fluorescence quenching microscopy (FQM) has enabled rapid, inexpensive, and high-fidelity visualization of two-dimensional (2D) materials such as graphene-based sheets and MoS2.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service