Skip to Content
Merck
All Photos(1)

Key Documents

359521

Sigma-Aldrich

(Aminomethyl)cyclopropane

97%

Synonym(s):

Cyclopropanemethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5CH2NH2
CAS Number:
Molecular Weight:
71.12
Beilstein:
773674
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

impurities

≤15% water

bp

86 °C/758 mmHg (lit.)

density

0.82 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

O.NCC1CC1

InChI

1S/C4H9N.H2O/c5-3-4-1-2-4;/h4H,1-3,5H2;1H2

InChI key

BIVIWWZEIMDEHV-UHFFFAOYSA-N

General description

Infrared (3500-50cm-1) spectra of gaseous and Raman (3500-50cm-1) spectra of liquid aminomethyl cyclopropane (cyclopropyl methylamine) has been reported. It was extracted by solid-phase micro extraction (SPME) from black rice grain.

Application

(Aminomethyl)cyclopropane may be used in chemical synthesis.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Changes in nutritional constituents, anthocyanins, and volatile compounds during the processing of black rice tea.
Wu L, et al.
Food Science and Biotechnology, 22(4), 917-923 (2013)
Conformational and structural studies of aminomethyl cyclopropane from temperature dependent FT-IR spectra of rare gas solutions and ab initio calculations.
Durig JR, et al.
The Journal of Physical Chemistry A, 107(39), 7713-7726 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service