Skip to Content
Merck
All Photos(1)

Key Documents

340243

Sigma-Aldrich

(S)-(−)-4-Methyl-4-(trichloromethyl)-2-oxetanone

98%

Synonym(s):

(S)-(−)-3-Hydroxy-3-methyl-4,4,4-trichlorobutyric β-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5Cl3O2
CAS Number:
Molecular Weight:
203.45
Beilstein:
5730853
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

98%

optical activity

[α]27/D −6.0°, c = 2 in ethanol

bp

120 °C/0.1 mmHg (lit.)

mp

43-44 °C (lit.)

SMILES string

C[C@]1(CC(=O)O1)C(Cl)(Cl)Cl

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service