Skip to Content
Merck
All Photos(1)

Documents

339962

Sigma-Aldrich

2-Hydroxy-4,6-dimethylpyrimidine hydrochloride

98%

Synonym(s):

4,6-Dimethyl-2-hydroxypyrimidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O · HCl
CAS Number:
Molecular Weight:
160.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

260 °C (dec.) (lit.)

SMILES string

Cl.Cc1cc(C)nc(O)n1

InChI

1S/C6H8N2O.ClH/c1-4-3-5(2)8-6(9)7-4;/h3H,1-2H3,(H,7,8,9);1H

InChI key

IFXXETYYSLJRNF-UHFFFAOYSA-N

Application

2-Hydroxy-4,6-dimethylpyrimidine hydrochloride was employed in the stereoselective synthesis of meso-N,N′-dicarbethoxy-2,4-diaminopentane and meso-2,4-diaminopentane.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective synthesis of meso-N, N'-dicarbethoxy-2, 4-diaminopentane and meso-2, 4-diaminopentane.
Hutchins RO and Maryanoff BE.
The Journal of Organic Chemistry, 37(11), 1829-1830 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service