33750
1,3:4,6-Di-O-benzylidene-D-mannitol
≥98.0% (sum of enantiomers, HPLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C20H22O6
CAS Number:
Molecular Weight:
358.39
Beilstein:
1627753
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Assay
≥98.0% (sum of enantiomers, HPLC)
optical activity
[α]20/D −8.4±0.3°, c = 1% in acetone
mp
192-195 °C
SMILES string
O[C@@H]1COC(O[C@H]1[C@@H]2OC(OC[C@H]2O)c3ccccc3)c4ccccc4
InChI
1S/C20H22O6/c21-15-11-23-19(13-7-3-1-4-8-13)25-17(15)18-16(22)12-24-20(26-18)14-9-5-2-6-10-14/h1-10,15-22H,11-12H2/t15-,16-,17-,18-,19?,20?/m1/s1
InChI key
NVEGGHPETXMRSV-NIJYPJQDSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Starting component for the synthesis of optically active glycerol derivatives with a minimum of protective group chemistry; Synthesis of heterocyclic 2,5-dimethanol derivatives
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A.V.R. Rao et al.
Journal of the Chemical Society. Chemical Communications, 1273-1273 (1988)
T.K.M. Shing
Journal of the Chemical Society. Chemical Communications, 262-262 (1987)
M.T. Reetz et al.
The Journal of Organic Chemistry, 50, 5434-5434 (1985)
T. Schubert et al.
Tetrahedron, 39, 2211-2211 (1983)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service