336653
Potassium heptafluorotantalate(V)
99%
Synonym(s):
Dipotassium heptafluorotantalate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
99%
form
powder
solubility
H2O: slightly soluble(lit.)
density
4.56 g/mL at 25 °C (lit.)
SMILES string
[K+].[K+].F[Ta--](F)(F)(F)(F)(F)F
InChI
1S/7FH.2K.Ta/h7*1H;;;/q;;;;;;;2*+1;+5/p-7
InChI key
DPXAUKPQZOAHGF-UHFFFAOYSA-G
Application
Used in the electrochemical synthesis of TaB2.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Proc. Electrochem. Soc., 93-9, 584-584 (1993)
Chem. Abstr., 122, 18820q-18820q (1995)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service