Skip to Content
Merck
All Photos(1)

Documents

334227

Sigma-Aldrich

4-Chloro-2-fluorophenol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)OH
CAS Number:
Molecular Weight:
146.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.536 (lit.)

bp

103-104 °C/50 mmHg (lit.)

mp

20 °C (lit.)

density

1.378 g/mL at 25 °C (lit.)

SMILES string

Oc1ccc(Cl)cc1F

InChI

1S/C6H4ClFO/c7-4-1-2-6(9)5(8)3-4/h1-3,9H

InChI key

ZKMUKBBWORLNLA-UHFFFAOYSA-N

General description

4-Chloro-2-fluorophenol undergoes reductive dechlorination to yield 2-fluorophenol by the sulfate-reducing consortium.

Application

4-Chloro-2-fluorophenol was used in the enzymatic preparation of 5-chloro-3-fluorocatechol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reductive dechlorination of halogenated phenols by a sulfate-reducing consortium.
Haggblom MM.
FEMS Microbiology Ecology, 26(1), 35-41 (1998)
Inna P Solyanikova et al.
Applied and environmental microbiology, 69(9), 5636-5642 (2003-09-06)
The present study describes the (19)F nuclear magnetic resonance analysis of the conversion of 3-halocatechols to lactones by purified chlorocatechol 1,2-dioxygenase (ClcA2), chloromuconate cycloisomerase (ClcB2), and chloromuconolactone dehalogenase (ClcF) from Rhodococcus opacus 1cp grown on 2-chlorophenol. The 3-halocatechol substrates were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service