Skip to Content
Merck
All Photos(1)

Documents

327964

Sigma-Aldrich

3-Hydroxy-1,2,3-benzotriazin-4(3H)-one

98%

Synonym(s):

3,4-Dihydro-3-hydroxy-4-oxo-1,2,3-benzotriazine, DHBT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O2
CAS Number:
Molecular Weight:
163.13
Beilstein:
609382
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

188-189 °C (dec.) (lit.)

SMILES string

ON1N=Nc2ccccc2C1=O

InChI

1S/C7H5N3O2/c11-7-5-3-1-2-4-6(5)8-9-10(7)12/h1-4,12H

InChI key

HJBLUNHMOKFZQX-UHFFFAOYSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service