Skip to Content
Merck
All Photos(1)

Key Documents

318353

Sigma-Aldrich

1-Cyclopentene-1,2-dicarboxylic anhydride

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O3
CAS Number:
Molecular Weight:
138.12
Beilstein:
123650
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

96%

form

solid

refractive index

n20/D 1.4978 (lit.)

bp

133-135 °C/10 mmHg (lit.)

mp

42-45 °C (lit.)

SMILES string

O=C1OC(=O)C2=C1CCC2

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(4)7(9)10-6/h1-3H2

InChI key

GBBPFLCLIBNHQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Valuable Diels-Alder dienophile, for example, in approach to the bicycloundecane system in taxanes.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 1587-1587 (1992)
Journal of the Chemical Society. Perkin Transactions 1, 2281-2281 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service