All Photos(1)
About This Item
Linear Formula:
CH3C10H6COCH3
CAS Number:
Molecular Weight:
184.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
98%
form
solid
mp
63-65 °C (lit.)
SMILES string
CC(=O)c1ccc2cc(C)ccc2c1
InChI
1S/C13H12O/c1-9-3-4-13-8-11(10(2)14)5-6-12(13)7-9/h3-8H,1-2H3
InChI key
SPAYGOAEIJHIDE-UHFFFAOYSA-N
General description
Condensation of 6′-methyl-2′-acetonaphthone (6-methyl-2-acetonaphthone) with various m- and p-substituted benzaldehydes under solvent free conditions to yield E-2-propen-1-ones has been reported.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis, characterization and spectral correlations in substituted styryl 6-methyl-2-naphthyl ketones.
Thirunarayanan, G.
Indian J. Chem. B, 46(9), 1511-1511 (2007)
Peter Ulrich et al.
The American journal of tropical medicine and hygiene, 91(4), 824-832 (2014-09-04)
Because of emerging resistance to existing drugs, new chemical classes of antimalarial drugs are urgently needed. We have rationally designed a library of compounds that were predicted to accumulate in the digestive vacuole and then decrystallize hemozoin by breaking the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service