Skip to Content
Merck
All Photos(1)

Key Documents

284122

Sigma-Aldrich

5-Cholesten-3β-ol-7-one

95%

Synonym(s):

3β-Hydroxy-5-cholesten-7-one, 7-Ketocholesterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

95%

SMILES string

C[C@]12CC[C@@]([C@](CC[C@H](O)C3)(C)C3=CC4=O)([H])[C@]4([H])[C@]1([H])CC[C@]2([H])[C@]([H])(C)CCCC(C)C

InChI

1S/C27H44O2/c1-17(2)7-6-8-18(3)21-9-10-22-25-23(12-14-27(21,22)5)26(4)13-11-20(28)15-19(26)16-24(25)29/h16-18,20-23,25,28H,6-15H2,1-5H3/t18-,20+,21-,22+,23+,25+,26+,27-/m1/s1

InChI key

YIKKMWSQVKJCOP-ABXCMAEBSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vesa M Olkkonen
Current opinion in lipidology, 23(5), 462-470 (2012-07-21)
To offer a comprehensive review on the roles that oxysterols synthesized or engulfed by macrophages, or oxysterol-binding proteins in these cells, play in the development and progression of atherosclerotic lesions. Oxysterols abundant within the plaque have the capacity to potentiate
Luisa Tesoriere et al.
The British journal of nutrition, 110(2), 230-240 (2012-12-12)
7-Ketocholesterol (7-KC)-induced apoptosis of macrophages is considered a key event in the development of human atheromas. In the present study, the effect of indicaxanthin (Ind), a bioactive pigment from cactus pear fruit, on 7-KC-induced apoptosis of human monocyte/macrophage THP-1 cells
Jiahn-Dar Huang et al.
Biochimica et biophysica acta, 1821(4), 637-646 (2012-02-22)
Sterculic acid is a cyclopropene fatty acid with numerous biological activities. In this study we demonstrate that sterculic acid is a potent inhibitor of endoplasmic reticulum (ER) stress and related inflammation caused by 7-ketocholesterol (7KCh). 7KCh is a highly toxic
Dmitri Svistounov et al.
PloS one, 7(9), e46134-e46134 (2012-10-03)
Fenestrations are transcellular pores in endothelial cells that facilitate transfer of substrates between blood and the extravascular compartment. In order to understand the regulation and formation of fenestrations, the relationship between membrane rafts and fenestrations was investigated in liver sinusoidal
Brigitta Buttari et al.
Biochemical pharmacology, 86(1), 130-137 (2013-04-25)
Macrophages, the major cellular components of atherosclerotic plaques, consist of two main subsets: the pro-inflammatory, M1 or classically activated macrophages, and the anti-inflammatory, M2 or alternatively activated macrophages. The molecular and cellular mechanisms that orchestrate the macrophage polarization and activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service