Skip to Content
Merck
All Photos(1)

Key Documents

27814

Sigma-Aldrich

(−)-α-Copaene

technical grade, ≥90% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
3196858
EC Number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

technical grade

Assay

≥90% (sum of enantiomers, GC)

optical activity

[α]20/D −0.4±0.2°, neat

refractive index

n20/D 1.490

bp

124 °C/15 mmHg (lit.)

density

0.910 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)[C@@H]1CC[C@]2(C)[C@H]3CC=C(C)[C@@H]2[C@@H]13

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teun Dekker et al.
Parasites & vectors, 4, 183-183 (2011-09-23)
Native mosquito repellent plants have a good potential for integrated mosquito control in local settings. Ocimum forskolei, Lamiaceae, is used in Eritrea as a spatial mosquito repellent inside houses, either through crushing fresh plants or burning dry plants. We verified
Nurhayat Tabanca et al.
Biomolecules, 10(5) (2020-05-02)
The Mediterranean fruit fly, Ceratitis capitata (Wiedemann) (Diptera: Tephritidae) poses a major threat to fruit and vegetable production in the United States and throughout the world. New attractants and detection methods could improve control strategies for this invasive pest. In
Miriam Frida Karlsson et al.
Chemoecology, 27(5), 187-198 (2017-09-26)
Insects locate their host plants using mainly visual and olfactory cues, generally of the exploited plant structure. However, when the resource is difficult to access, it could be beneficial to utilise indirect cues, which indicates the presence of reward (e.g.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service