Skip to Content
Merck
All Photos(1)

Documents

275395

Sigma-Aldrich

4-Mercaptophenol

technical grade, 90%

Synonym(s):

4-Hydroxythiophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSC6H4OH
CAS Number:
Molecular Weight:
126.18
Beilstein:
2039306
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

bp

149-150 °C/25 mmHg (lit.)

mp

33-35 °C (lit.)

SMILES string

Oc1ccc(S)cc1

InChI

1S/C6H6OS/c7-5-1-3-6(8)4-2-5/h1-4,7-8H

InChI key

BXAVKNRWVKUTLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The density function theory calculations were performed to infer the IR and Raman spectra for 4-mercaptophenol molecule.

Application

4-Mercaptophenol was used as a resin monomer in the study of the different gold nanocrystal core-resin shell structures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Seunghwan Lee et al.
Dalton transactions (Cambridge, England : 2003), 50(28), 9958-9967 (2021-07-07)
Area selective atomic layer deposition (AS-ALD) is a promising future technology for the realization of a 5 nm scale Si complementary field effect transistor (FET) and its application in industry. AS-ALD is one of the "bottom-up" technologies, which is a
Zhenhua Sun et al.
Langmuir : the ACS journal of surfaces and colloids, 28(24), 9082-9092 (2012-03-22)
The formation of different Au nanocrystal core-resin shell structures through the control of the nanocrystal assembly and shell polymerization is investigated. 4-Mercaptophenol is employed together with formaldehyde as the resin monomers. 4-Mercaptophenol molecules bond to the surface of Au nanocrystals
Ran Li et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 122, 698-703 (2013-12-24)
In this paper, 4-mercaptophenol (4-MPH) was designed as a model molecule for theoretical and experimental studies of the molecule structure. Density functional theory (DFT) calculations have been performed to predict the IR and Raman spectra for the molecule. In addition
Isabel Freitag et al.
Journal of biomedical optics, 20(5), 55002-55002 (2015-05-06)
Identification of tumor and normal cells is a promising application of Raman spectroscopy. The throughput of Raman-assisted cell sorting is limited by low sensitivity. Surface-enhanced Raman spectroscopy (SERS) is a well-recognized candidate to increase the intensity of Raman signals of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service