Skip to Content
Merck
All Photos(1)

Key Documents

264911

Sigma-Aldrich

7-Methylquinoline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N
CAS Number:
Molecular Weight:
143.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
form:
crystals
Assay:
97%
Pricing and availability is not currently available.

Assay

97%

form

crystals

bp

258 °C (lit.)

mp

35-37 °C (lit.)

SMILES string

Cc1ccc2cccnc2c1

InChI

1S/C10H9N/c1-8-4-5-9-3-2-6-11-10(9)7-8/h2-7H,1H3

InChI key

KDYVCOSVYOSHOL-UHFFFAOYSA-N

General description

7-methylquinoline has been bioassayed as tumor initiator on the skin of Sencar mice[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

88. The Skraup reaction with m-substituted anilines.
Bradford L, et al.
Journal of the Chemical Society, 437-445 (1947)
E J La Voie et al.
Carcinogenesis, 4(9), 1169-1173 (1983-09-01)
Quinoline is a hepatocarcinogen in mice and rats, a mutagen in Salmonella typhimurium, and induces unscheduled DNA synthesis in primary cultures of rat hepatocytes. In contrast, isoquinoline has not been shown to be genotoxic. The metabolites of quinoline and isoquinoline

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service