Skip to Content
Merck
All Photos(1)

Key Documents

261157

Sigma-Aldrich

Neodymium

ingot (in mineral oil), 99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nd
CAS Number:
Molecular Weight:
144.24
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.9% trace rare earth metals basis

form

ingot (in mineral oil)

reaction suitability

reagent type: catalyst
core: neodymium

resistivity

64.0 μΩ-cm, 20°C

bp

3074 °C (lit.)

mp

1021 °C (lit.)

density

7.003 g/mL at 25 °C (lit.)

SMILES string

[Nd]

InChI

1S/Nd

InChI key

QEFYFXOXNSNQGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angie Phelan et al.
Australian orthodontic journal, 28(2), 159-169 (2013-01-12)
The demand for clear sequential aligner therapy has increased dramatically in recent years. An improved system utilising small neodymium-iron-boron (NdFeB) magnetic attachments has been proposed to enhance appliance capabilities. The aim of the investigation was to analyse the force system
Ionut Mihalcea et al.
Inorganic chemistry, 51(18), 9610-9618 (2012-08-31)
Three series of mixed uranyl-lanthanide (Ce or Nd) carboxylate coordination polymers have been successfully synthesized by means of a hydrothermal route using either conventional or microwave heating methods. These compounds have been prepared from mixtures of uranyl nitrate, lanthanide nitrate
Haohai Yu et al.
Scientific reports, 3, 1085-1085 (2013-01-22)
Two-dimensional (2D) visible crystal-structure patterns analogous to the quantum harmonic oscillator (QHO) have been experimentally observed in the near- and far-fields of a self-frequency-doubling (SFD) microchip laser. Different with the fundamental modes, the localization of the SFD light is changed
Anuja Bokare et al.
Colloids and surfaces. B, Biointerfaces, 102, 273-280 (2012-09-27)
Nanosized (8-9 nm) Nd doped and Ag coated TiO(2) nanoparticles have been synthesized by sol-gel method. The physicochemical properties of these particles were investigated by X-ray diffraction (XRD), diffuse reflectance UV-visible (DRUV) spectra and Brunauer-Emmett-Teller (BET) surface area analysis. The
Guillaume Soulet et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(17), 6682-6687 (2013-04-10)
Continental ice sheets are a key component of the Earth's climate system, but their internal dynamics need to be further studied. Since the last deglaciation, the northern Eurasian Fennoscandian Ice Sheet (FIS) has been connected to the Black Sea (BS)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service