Skip to Content
Merck
All Photos(1)

Key Documents

259039

Sigma-Aldrich

Chloro(1,5-hexadiene)rhodium(I),dimer

98%

Synonym(s):

Di-μ-chlorobis[(1,2,5,6-η)-1,5-hexadiene]dirhodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(H2C=CHCH2CH2CH=CH2)RhCl]2
CAS Number:
Molecular Weight:
434.96
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

core: rhodium
reagent type: catalyst

mp

118-120 °C (lit.)

SMILES string

Cl[Rh].Cl[Rh].C=CCCC=C.C=CCCC=C

InChI

1S/2C6H10.2ClH.2Rh/c2*1-3-5-6-4-2;;;;/h2*3-4H,1-2,5-6H2;2*1H;;/q;;;;2*+1/p-2

InChI key

HXNWPVBAYNUADR-UHFFFAOYSA-L

Application

Catalyst for:
  • Hydrogenation
  • Enantioselective hydrosilylation
Useful catalyst for the reduction of arenes and heterocyclic compounds using CTAB or THS as phase-transfer catalyst.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 18, 111-111 (1985)
Organometallics, 2, 1055-1055 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service