Skip to Content
Merck
All Photos(2)

Key Documents

249408

Sigma-Aldrich

2,6-Difluorobenzoyl chloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3COCl
CAS Number:
Molecular Weight:
176.55
Beilstein:
639438
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

99%

form

liquid

refractive index

n20/D 1.501 (lit.)

bp

72-77 °C/13 mmHg (lit.)

density

1.404 g/mL at 25 °C (lit.)

SMILES string

Fc1cccc(F)c1C(Cl)=O

InChI

1S/C7H3ClF2O/c8-7(11)6-4(9)2-1-3-5(6)10/h1-3H

InChI key

QRHUZEVERIHEPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Difluorobenzoyl chloride has been used in:
  • the preparation of series of novel 2-aryl substituted 4H-3,1-benzoxazin-4-ones[1]
  • regioselective synthesis of 3,6-disubstituted-2-aminoimidazo[1,2-a]pyridine[2]
  • Friedel-Crafts acylation reaction of toluene, anisol, thioanisol, 4-phenoxyacetophenone and N,N-diacetyl-4-phenoxyaniline[3]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Jakobsen et al.
Bioorganic & medicinal chemistry, 8(8), 2095-2103 (2000-09-26)
The synthesis of a series of novel 2-aryl substituted 4H-3,1-benzoxazin-4-ones and their evaluation as specific inhibitors of the Tissue Factor (TF)/Factor VIIa (FVIIa)-induced pathway of coagulation is reported. Inhibitory activities (IC50 values) in the range 0.17 to > 40 microM
Regioselective synthesis of 3, 6-disubstituted-2-aminoimidazo [1, 2-a] pyridines.
Jaramillo C, et al.
Tetrahedron Letters, 43(50), 9051-9054 (2002)
New polymer syntheses, 36. Functionalized aromatic polyethers derived from 4'-substituted 2, 6-difluorobenzophenones.
Kricheldorf HR and Delius U.
Makromol. Chem., 190(6), 1277-1288 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service