244139
Magnesium chloride
powder, −325 mesh, 98%
Synonym(s):
Magnogene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98%
form
powder
impurities
<15%
particle size
−325 mesh
mp
714 °C (lit.)
density
2.32 g/mL at 25 °C (lit.)
SMILES string
Cl[Mg]Cl
InChI
1S/2ClH.Mg/h2*1H;/q;;+2/p-2
InChI key
TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
Useful catalyst for the selective oxidative coupling of methane.
Other Notes
Contains a trace amount of insoluble magnesium
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Applied Catalysis, 50, 223-223 (1989)
Scientific reports, 5, 9919-9919 (2015-06-06)
We report a biosensing platform for viral load measurement through electrical sensing of viruses on a flexible plastic microchip with printed electrodes. Point-of-care (POC) viral load measurement is of paramount importance with significant impact on a broad range of applications
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service