Skip to Content
Merck
All Photos(1)

Key Documents

239933

Sigma-Aldrich

Butyronitrile

≥99%

Synonym(s):

Propyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CN
CAS Number:
Molecular Weight:
69.11
Beilstein:
1361452
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

2.4 (vs air)

vapor pressure

23 mmHg ( 25 °C)

Assay

≥99%

autoignition temp.

910 °F

refractive index

n20/D 1.384 (lit.)

bp

115-117 °C (lit.)

mp

−112 °C (lit.)

density

0.794 g/mL at 25 °C (lit.)

SMILES string

CCCC#N

InChI

1S/C4H7N/c1-2-3-4-5/h2-3H2,1H3

InChI key

KVNRLNFWIYMESJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E G DeMaster et al.
Biochemical pharmacology, 46(1), 117-123 (1993-07-06)
n-Butyraldoxime (n-BO) is known to cause a disulfiram/ethanol-like reaction in humans, a manifestation of the inhibition of hepatic aldehyde dehydrogenase (AIDH). As with a number of other in vivo inhibitors of AIDH, n-BO does not inhibit purified AIDH in vitro
T D Thomas et al.
Physical chemistry chemical physics : PCCP, 9(6), 719-724 (2007-02-03)
Experimental evidence for an effect of molecular conformation on inner-shell ionization energies has been observed for the first time. Examples are seen in the carbon 1s spectra of butyronitrile, 1-fluoropropane, and propanal, and other similar molecules. At room temperature these
G Grampp et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 58(6), 1219-1226 (2002-05-08)
Dimerization enthalpies and equilibrium constants have been determined for the radical anion of 2,3-dichloro-5,6-dicyano-p-benzoquinone (DDQ), the radical cations of N,N,N',N'-tetramethyl-p-phenylenediamine, N,N-dimethyl-p-phenylenediamine, 2,3,5,6-tetramethyl-p-phenylenediamine, N,N,N',N'-tetraethyl-p-phenylenediamine, N,N-diethyl-p-phenylenediamine and N,N,N'-trimethyl-p-phenylenediamine. Neutral radicals investigated are 2,2,6,6-tetramethylpiperidine-1-oxyl (TEMPO) and galvinoxyl. Solvent used was acetone, EtOH/Et2O-mixture (2:1
J M Samet et al.
Analytical biochemistry, 182(1), 32-36 (1989-10-01)
An HPLC method for the separation and analysis of arachidonic acid and eight phospholipid classes is described: phosphatidylglycerol, phosphatidylinositol, cardiolipin, phosphatidylserine, phosphatidylethanolamine, phosphatidylcholine, sphingomyelin, and 2-lysophosphatidylcholine. The separation is carried out at 60 degrees C on 2 cyanopropyl columns using
M S Nawaz et al.
Applied and environmental microbiology, 58(1), 27-31 (1992-01-01)
A strain of Klebsiella pneumoniae that used aliphatic nitriles as the sole source of nitrogen was adapted to benzonitrile as the sole source of carbon and nitrogen. Gas chromatographic and mass spectral analyses of culture filtrates indicated that K. pneumoniae

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service