Skip to Content
Merck
All Photos(1)

Key Documents

235911

Sigma-Aldrich

Tetraethylammonium fluoride hydrate

98%

Synonym(s):

Tetraethylammonium fluoride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4NF·xH2O
CAS Number:
Molecular Weight:
149.25 (anhydrous basis)
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
98%
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

solid

functional group

amine

SMILES string

O.[F-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.FH.H2O/c1-5-9(6-2,7-3)8-4;;/h5-8H2,1-4H3;1H;1H2/q+1;;/p-1

InChI key

YEKUWOWHPVKTCQ-UHFFFAOYSA-M

General description

Tetraethylammonium fluoride hydrate is a quaternary ammonium salt, is commonly used as a deprotection reagent in organic synthesis for the deprotection of silyl ether functional groups. It also acts as a mild and selective fluoride ion donor in fluorination reactions.

Application

Tetraethylammonium fluoride hydrate can be used as a homogeneous catalyst in the selective methanolytic depolymerization of poly(lactic acid) PLA.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun Jiao et al.
Dalton transactions (Cambridge, England : 2003), 46(7), 2270-2280 (2017-01-31)
Many synthesis parameters can influence zeolite crystallization, which include the molar ratio of reagents, water content, temperature, the selection of extraframework cations (organic or inorganic template) and so on. In this paper, two new materials, BUCT-1 with cuboid morphology and
Frank C Hendriks et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(26), 6305-6314 (2017-02-22)
A micro-spectroscopic method has been developed to probe the accessibility of zeolite crystals using a series of fluorescent 4-(4-diethylaminostyryl)-1-methylpyridinium iodide (DAMPI) probes of increasing molecular size. Staining large zeolite crystals with MFI (ZSM-5) topology and subsequent mapping of the resulting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service