Skip to Content
Merck
All Photos(1)

Key Documents

235245

Sigma-Aldrich

4-Chlorophenyl dichlorophosphate

99%

Synonym(s):

4-Chlorophenyl phosphorodichloridate, 4-Chlorophenyl dichlorophosphinate, p-Chlorophenyl dichlorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2P(O)OC6H4Cl
CAS Number:
Molecular Weight:
245.43
Beilstein:
910754
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

99%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

refractive index

n20/D 1.539 (lit.)

bp

142 °C/11 mmHg (lit.)

density

1.508 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(OP(Cl)(Cl)=O)cc1

InChI

1S/C6H4Cl3O2P/c7-5-1-3-6(4-2-5)11-12(8,9)10/h1-4H

InChI key

CCZMQYGSXWZFKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Antimicrobial agents
  • Antioxidant agents
  • Antitumor agents
  • Nucleotide anti-viral prodrug

Used as:
  • Phosphorylation agent

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service