Skip to Content
Merck
All Photos(1)

Documents

224995

Sigma-Aldrich

Methyl 2-methyl-3-furancarboxylate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.473 (lit.)

bp

75 °C/20 mmHg (lit.)

density

1.116 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccoc1C

InChI

1S/C7H8O3/c1-5-6(3-4-10-5)7(8)9-2/h3-4H,1-2H3

InChI key

UVRRIABXNIGUJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 2-methyl-3-furancarboxylate was used in the synthesis of sulfonylfuran ureas.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krista B Goodman et al.
Bioorganic & medicinal chemistry letters, 19(1), 27-30 (2008-12-09)
Endothelial lipase (EL) activity has been implicated in HDL catabolism, vascular inflammation, and atherogenesis, and inhibitors are therefore expected to be useful for the treatment of cardiovascular disease. Sulfonylfuran urea 1 was identified in a high-throughput screening campaign as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service