209724
Glycolaldehyde diethyl acetal
98%
Synonym(s):
2,2-Diethoxyethanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H14O3
CAS Number:
Molecular Weight:
134.17
MDL number:
PubChem Substance ID:
Recommended Products
Assay
98%
SMILES string
CCOC(CO)OCC
InChI
1S/C6H14O3/c1-3-8-6(5-7)9-4-2/h6-7H,3-5H2,1-2H3
InChI key
IKKUKDZKIIIKJK-UHFFFAOYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service