Skip to Content
Merck
All Photos(1)

Key Documents

20646

Sigma-Aldrich

O-tert-Butyl-L-threonine tert-butyl ester acetate salt

≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25NO3 · C2H4O2
CAS Number:
Molecular Weight:
291.38
Beilstein:
6707078
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

optical activity

[α]20/D −10±1°, c = 1% in methanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

59-61 °C

application(s)

peptide synthesis

SMILES string

CC(O)=O.C[C@@H](OC(C)(C)C)[C@H](N)C(=O)OC(C)(C)C

InChI

1S/C12H25NO3.C2H4O2/c1-8(15-11(2,3)4)9(13)10(14)16-12(5,6)7;1-2(3)4/h8-9H,13H2,1-7H3;1H3,(H,3,4)/t8-,9+;/m1./s1

InChI key

BGAUVMFJRASONL-RJUBDTSPSA-N

Other Notes

Protected threonine for peptide synthesis, e.g. of glycopeptides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chernyak AYa et al.
Carbohydrate research, 216, 381-398 (1991-09-02)
The disaccharides alpha-L-Rhap-(1----3)-beta-D-GlcpA and beta-D-GlcpA-(1----3)-alpha-L-Rhap bearing amide-linked L-serine or L-threonine, which represent the repeating unit(s) of the capsular polysaccharide from E. coli O6:K54:H10, have been synthesised. O-tert-Butyl-protected amino acid tert-butyl esters were condensed with the corresponding biouronic acid as the
N.J. Forrow et al.
Tetrahedron Letters, 31, 3493-3493 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service