Skip to Content
Merck
All Photos(1)

Documents

199028

Sigma-Aldrich

2-(3-Hydroxyphenyl)ethanol

99%

Synonym(s):

3-Hydroxyphenethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CH2OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
2433753
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.564 (lit.)

bp

168-173 °C/4 mmHg (lit.)

density

1.082 g/mL at 25 °C (lit.)

SMILES string

OCCc1cccc(O)c1

InChI

1S/C8H10O2/c9-5-4-7-2-1-3-8(10)6-7/h1-3,6,9-10H,4-5H2

InChI key

AMQIPHZFLIDOCB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The inelastic electron tunneling (IET) spectra of 2-(3-hydroxyphenyl)ethanol was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rocío Casadey et al.
Food chemistry, 285, 275-281 (2019-02-25)
The exposure of fatty products to environmental light can trigger lipid oxidation in food through a sensitized-photooxidation process, which involves the participation of the species singlet oxygen (O2(1Δg)). Therefore, preservation of food quality represents a subject of great economic interest
Rocío Moreno-González et al.
Journal of chromatography. A, 1609, 460434-460434 (2019-08-17)
Table olives contain a wide range of polyphenols responsible for protective effects on health that have been associated with a lower prevalence of chronic diseases. A new method to identify and quantify these compounds in table olives, by means of
Min Kyung Song et al.
Journal of agricultural and food chemistry, 67(7), 2028-2035 (2019-01-31)
Caffeic acid phenethyl ester (CAPE) is an ester of a hydroxycinnamic acid (phenylpropanoid) and a phenylethanoid (2-phenylethanol; 2-PE), which has long been used in traditional medicine. Here, we synthesized 54 hydroxycinnamic acid-phenylethanoid esters by feeding 64 combinations of hydroxycinnamic acids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service