Skip to Content
Merck
All Photos(1)

Key Documents

195863

Sigma-Aldrich

2,3,4-Trimethoxybenzyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CH2OH
CAS Number:
Molecular Weight:
198.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.532 (lit.)

bp

105 °C/25 mmHg (lit.)

density

1.151 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(CO)c(OC)c1OC

InChI

1S/C10H14O4/c1-12-8-5-4-7(6-11)9(13-2)10(8)14-3/h4-5,11H,6H2,1-3H3

InChI key

DGJVVEVPKPOLEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Photoelectrocatalytic partial oxidation of 2,3,4-trimethoxybenzyl alcohol in aqueous solution irradiated by near-UV light has been investigated in three-electrode batch reactor.

Application

2,3,4-Trimethoxybenzyl alcohol was used to introduce a benzyl group onto the 2′-OH of purine ribonucleoside.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigetada Kozai et al.
Nucleosides, nucleotides & nucleic acids, 22(2), 145-151 (2003-05-15)
A new method to introduce a benzyl group onto the 2'-OH of purine ribonucleoside is described. Thus, 6-chloropurine 3'-O-benzoylriboside and its 5'-O-trityl congener were condensed with benzyl alcohol using the Mitsunobu reaction to give the 2'-O-benzyl derivative. The yields were
Photoelectrocatalytic selective oxidation of 4-methoxybenzyl alcohol in water by TiO2 supported on titanium anodes.
Ozcan L, et al.
Applied Catalysis. B, Environmental, 132, 535-542 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service