Skip to Content
Merck
All Photos(2)

Documents

195405

Sigma-Aldrich

3-(4-Methylphenoxy)benzaldehyde

97%

Synonym(s):

3-(p-Tolyloxy)benzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OC6H4CHO
CAS Number:
Molecular Weight:
212.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.59 (lit.)

bp

140 °C/2 mmHg (lit.)

density

1.102 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(Oc2cccc(C=O)c2)cc1

InChI

1S/C14H12O2/c1-11-5-7-13(8-6-11)16-14-4-2-3-12(9-14)10-15/h2-10H,1H3

InChI key

ASKLCRGXIJGVOY-UHFFFAOYSA-N

Application

3-(4-Methylphenoxy)benzaldehyde was used in the synthesis of benzoxazole derivatives of the mannopeptimycin glycopeptide antibiotics.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phaik-Eng Sum et al.
Bioorganic & medicinal chemistry letters, 13(15), 2607-2610 (2003-07-11)
A series of benzoxazole derivatives of the mannopeptimycin glycopeptide antibiotics was synthesized via a novel benzoxazole formation reaction by treating aminophenol of mannopeptimycin-beta with an aldehyde and DDQ in DMF. Some of these derivatives (e.g., 5b, 5d, 5m, and 7b)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service