Skip to Content
Merck
All Photos(1)

Key Documents

192651

Sigma-Aldrich

Furoin

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

solid

mp

134-137 °C (lit.)

solubility

acetone: 0.25 g/10 mL, clear, colorless to deep brown-yellow

SMILES string

OC(c1ccco1)C(=O)c2ccco2

InChI

1S/C10H8O4/c11-9(7-3-1-5-13-7)10(12)8-4-2-6-14-8/h1-6,9,11H

InChI key

MIJRFWVFNKQQDK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Furoin was used as fluorogenic reagent for the selective and sensitive liquid chromatographic determination of various guanidines[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arthur L Weber et al.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life, 49(4), 199-211 (2019-12-10)
The chemistry of imidazolium-catalyzed imidazolium synthesis was studied as part of an effort to develop a plausible prebiotic synthesis of a small catalytic molecule capable of catalyzing its own synthesis. Specifically, we investigated the one-pot 1-ethyl-3-methylimidazolium acetate (EMIM-Ac) catalyzed synthesis
Jun-Hui Zhang et al.
Analytica chimica acta, 999, 169-175 (2017-12-20)
Porous organic molecular cages, as a new type of porous materials, have in recent years attracted a tremendous amount of attention for their potential applications. However, to the best of our knowledge, there has been no attempt to utilize porous
R Gatti et al.
Journal of pharmaceutical and biomedical analysis, 48(3), 754-759 (2008-09-27)
Furoin, a benzoin analogue, was examined as novel fluorogenic reagent for the selective and sensitive LC determination of various guanidines after pre-column derivatization. The derivatization reaction was carried out at 100 degrees C for 5 min to give adducts that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service