Skip to Content
Merck
All Photos(1)

Key Documents

192112

Sigma-Aldrich

Borane dimethyl sulfide complex solution

2.0 M in diethyl ether

Synonym(s):

BMS, Trihydro[thiobis[methane]]boron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S · BH3
CAS Number:
Molecular Weight:
75.97
Beilstein:
3663489
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

form

liquid

reaction suitability

reagent type: reductant

concentration

2.0 M in diethyl ether

density

0.74 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

B.CSC

InChI

1S/C2H6S.BH3/c1-3-2;/h1-2H3;1H3

InChI key

RMHDLBZYPISZOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant used as a regioselective reducing agent

Reactant involved in:
  • Hydroboration / oxidation
Synthesis of cage compounds

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Repr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saskia Ditisheim et al.
Inflammatory bowel diseases, 21(11), 2598-2604 (2015-08-06)
Inflammatory bowel diseases (IBD) are systemic conditions that commonly display extraintestinal manifestations. Inflammatory articular disease (IAD: axial or peripheral) is the most common extraintestinal manifestation. The aim of this study was to evaluate the prevalence and the clinical characteristics associated
Isidoro González-Álvaro et al.
Rheumatology (Oxford, England), 54(7), 1200-1209 (2014-12-21)
The aim of this study was to establish guidelines for the optimization of biologic therapies for health professionals involved in the management of patients with RA, AS and PsA. Recommendations were established via consensus by a panel of experts in
Elin S Gray et al.
Oncotarget, 6(39), 42008-42018 (2015-11-03)
Repeat tumor biopsies to study genomic changes during therapy are difficult, invasive and data are confounded by tumoral heterogeneity. The analysis of circulating tumor DNA (ctDNA) can provide a non-invasive approach to assess prognosis and the genetic evolution of tumors
Jeong Won Jang et al.
Hepatology (Baltimore, Md.), 61(6), 1809-1820 (2015-01-30)
The effect of viral suppression on long-term disease outcome after decompensation in patients with hepatitis B virus (HBV)-related cirrhosis has not been established. The aim of this study was to determine the long-term effect of antiviral therapy (AVT) in patients
Veena K Ranganath et al.
Arthritis care & research, 67(7), 929-939 (2015-01-13)
To evaluate the effect of sustained American College of Rheumatology (ACR)/European League Against Rheumatism (EULAR) Boolean remission on residual joint inflammation assessed by magnetic resonance imaging (MRI) and to secondarily evaluate other clinical definitions of remission, within an early seropositive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service