Skip to Content
Merck
All Photos(1)

Key Documents

187410

Sigma-Aldrich

5-Methyl-2-nitrobenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CH2OH
CAS Number:
Molecular Weight:
167.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

mp

66-67 °C (lit.)

functional group

hydroxyl
nitro

SMILES string

Cc1ccc(c(CO)c1)[N+]([O-])=O

InChI

1S/C8H9NO3/c1-6-2-3-8(9(11)12)7(4-6)5-10/h2-4,10H,5H2,1H3

InChI key

IKEYTRGLCHZQHO-UHFFFAOYSA-N

General description

5-Methyl-2-nitrobenzyl alcohol undergoes oxidation in the presence of ammonium chlorochromate adsorbed on montmorillonite K-10 to yield 5-methyl-2-nitrobenzaldehyde[1]. It also undergoes oxidation with chromium trioxide-HZSM-5 zeolite under microwave irradiation on solventless system[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammonium chlorochromate adsorbed on montmorillonite K-10: Selective oxidation of alcohols under solvent-free conditions.
Heravi M M and Hydar KT.
J. Chem. Res. Synop., 10, 656-657 (1998)
Remarkable fast microwave-assisted zeolite HZSM-5 catalyzed oxidation of alcohols with chromium trioxide under solvent-free conditions.
Heravi MM.
J. Chem. Res. Synop., 5, 334-335 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service