Skip to Content
Merck
All Photos(1)

Key Documents

166979

Sigma-Aldrich

Dimethyl 3-hydroxyglutarate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH2CH(OH)CH2COOCH3
CAS Number:
Molecular Weight:
176.17
Beilstein:
1776666
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.442 (lit.)

bp

139-140 °C/8 mmHg (lit.)

density

1.192 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CC(O)CC(=O)OC

InChI

1S/C7H12O5/c1-11-6(9)3-5(8)4-7(10)12-2/h5,8H,3-4H2,1-2H3

InChI key

CUPGMRSSZADEIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Enantioselective hydrolysis and ammonolysis of dimethyl-3-hydroxyglutarate has been studied using immobilized lipase B isolated from Candida antarctica.

Application

Dimethyl 3-hydroxyglutarate was used in the synthesis of two chiral fragments representing C1-11 and C12-25 of the polyene macrolide pimaricin.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective enzymatic preparation of chiral glutaric monocarboxylic acids and amides.
Jacobsen EE, et al.
Journal of Molecular Catalysis. B, Enzymatic, 21(1), 55-58 (2003)
A convergent synthetic strategy for the polyene macrolide pimaricin.
Brooks DW and Palmer JT.
Tetrahedron Letters, 24(30), 3059-3062 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service