15545
Boc-Arg(Mts)-OH cyclohexylammonium salt
≥99.0% (TLC)
Synonym(s):
Nα-Boc-Nω-(mesitylene-2-sulfonyl)-L-arginine cyclohexylammonium salt
About This Item
Recommended Products
Assay
≥99.0% (TLC)
form
solid
optical activity
[α]20/D +11.5±1°, c = 0.5% in methanol
reaction suitability
reaction type: Boc solid-phase peptide synthesis
mp
~130 °C
application(s)
peptide synthesis
SMILES string
NC1CCCCC1.Cc2cc(C)c(c(C)c2)S(=O)(=O)NC(=N)NCCC[C@@H](NC(=O)OC(C)(C)C)C(O)=O
InChI
1S/C20H32N4O6S.C6H13N/c1-12-10-13(2)16(14(3)11-12)31(28,29)24-18(21)22-9-7-8-15(17(25)26)23-19(27)30-20(4,5)6;7-6-4-2-1-3-5-6/h10-11,15H,7-9H2,1-6H3,(H,23,27)(H,25,26)(H3,21,22,24);6H,1-5,7H2/t15-;/m1./s1
InChI key
YZLIRHMRDOFTHQ-XFULWGLBSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service