Skip to Content
Merck
All Photos(1)

Key Documents

149853

Sigma-Aldrich

1-(α,α,α-Trifluoro-m-tolyl)piperazine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13F3N2
CAS Number:
Molecular Weight:
230.23
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

96%

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

refractive index

n20/D 1.521 (lit.)

density

1.226 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1cccc(c1)N2CCNCC2

InChI

1S/C11H13F3N2/c12-11(13,14)9-2-1-3-10(8-9)16-6-4-15-5-7-16/h1-3,8,15H,4-7H2

InChI key

KKIMDKMETPPURN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Serotonin receptor agonist with preference for 5-HT1B serotonin receptors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Zwartsen et al.
Toxicology and applied pharmacology, 397, 115015-115015 (2020-04-23)
Hyperthermia is a well-known, potentially life-threatening, side effect of stimulant psychoactive substances that worsens the neurological outcome of hospitalized patients. However, current in vitro methods to assess the hazard of psychoactive substances do not account for hyperthermia. Therefore, this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service