Skip to Content
Merck
All Photos(1)

Key Documents

144614

Sigma-Aldrich

4-Chlorocinnamonitrile, mixture of cis and trans

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH=CHCN
CAS Number:
Molecular Weight:
163.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

solid

mp

55-70 °C (lit.)

functional group

chloro
nitrile

SMILES string

Clc1ccc(\C=C\C#N)cc1

InChI

1S/C9H6ClN/c10-9-5-3-8(4-6-9)2-1-7-11/h1-6H/b2-1+

InChI key

PPCNBCKABHGVMX-OWOJBTEDSA-N

Application

4-Chlorocinnamonitrile was used in the synthesis of ethyl-1-(4-chlorophenyl)-2-cyanoethyl(diethoxymethyl)phosphinate[1]. It was used to compare the radical and homocopolymerization ability of geometrical isomers of cinnamonitriles[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diethoxymethylphosphonites and phosphinates. Intermediates for thesynthesis of a, ?-and X aminoalkylphosphonous acids.
Dingwalla JG, et al.
Tetrahedron, 45(12), 3787-3808 (1989)
Radical homo-and copolymerization of ring-substituted trans-and cis-cinnamonitriles.
Tanaka H, et al.
Makromol. Chem., Rapid Commun., 14(10), 649-655 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service