Skip to Content
Merck
All Photos(1)

Key Documents

123099

Sigma-Aldrich

3-Morpholinopropylamine

98%

Synonym(s):

4-(3-Aminopropyl)morpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16N2O
CAS Number:
Molecular Weight:
144.21
Beilstein:
105104
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.4761 (lit.)

bp

224 °C (lit.)

mp

−15 °C (lit.)

density

0.987 g/mL at 25 °C (lit.)

SMILES string

NCCCN1CCOCC1

InChI

1S/C7H16N2O/c8-2-1-3-9-4-6-10-7-5-9/h1-8H2

InChI key

UIKUBYKUYUSRSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service