Skip to Content
Merck
All Photos(1)

Key Documents

117471

Sigma-Aldrich

N-tert-Butylformamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONHC(CH3)3
CAS Number:
Molecular Weight:
101.15
Beilstein:
1738869
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Assay

98%

refractive index

n20/D 1.433 (lit.)

bp

202 °C (lit.)

mp

16 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

functional group

amide

SMILES string

CC(C)(C)NC=O

InChI

1S/C5H11NO/c1-5(2,3)6-4-7/h4H,1-3H3,(H,6,7)

InChI key

SDLAKRCBYGZJRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-tert-Butylformamide forms nanostructure due to segregation of the molecules into polar and non-polar domains, analogous to amphiphile self-assembly.

Application

N-tert-Butylformamide was used to investigate the ability of low molecular weight amides to function as non-aqueous amphiphile self-assembly media.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamar L Greaves et al.
Physical chemistry chemical physics : PCCP, 13(20), 9180-9186 (2011-04-07)
The ability of low molecular weight amides to support amphiphile self-assembly is shown to be a general feature for this class of solvents. This report extends the number of known polar solvents which can support amphiphile self-assembly by five new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service